Order Entry
United States
ContactUsLinkComponent
Ethidium bromide 10 mg/mL in aqueous solution for molecular biology
Toxic
Health
Ethidium bromide 10 mg/mL in aqueous solution for molecular biology
Catalog # IB40075
Supplier:  IBI Scientific
CAS Number:  
Toxic
Health
Ethidium bromide 10 mg/mL in aqueous solution for molecular biology
Catalog # IB40075
Supplier:  IBI Scientific
Supplier Number:  IB40075
CAS Number:  

Specifications

  • Pack type:
    Dropping bottle
  • Size:
    10 mL
  • Cat. No.:
    IB40075
  • Supplier no.:
    40075

Specifications

About this item

 :
 :394.31 g/mol
EINECS:214-984-6
 :GHS06
 :GHS08
CAS Number:1239-45-8
Formula:C₂₁H₂₀BrN₃
MDL Number:MFCD00011724