
Ethanol denatured 89.5-91.5%, OmniSolv® for HPLC, for LC-MS, for spectrophotometry (denatured with 5% isopropanol and 5% methanol), Supelco®
:64-17-5
:
:H₃CCH₂OH
:MFCD00003568
:46.07 g/mol
:Ambient
:200-578-6
:1170
:3,II
:GHS02
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:EPA Safer chemical solvent (green circle rating); Chem21 solvent selection guide recommended by default and after discussion
- Assay (Ethanol):89.5-91.5%
- Assay (Isopropyl alcohol):4.0-6.0%
- Assay (Methanol):3.5-5.5%
- Color (APHA):10 max
- Filtered through 0.2 µm filter:To pass test
- Form:Clear liquid
- Infrared spectrum:Conforms to standard
- Residue after evaporation:4ppm max
- Titrable acid:0.3µeq/g max
- Titrable base:0.04µeq/g max
- UV Abs. at 205 nm:1.00AU max
- UV Abs. at 210 nm:0.65AU max
- UV Abs. at 220 nm:0.35AU max
- UV Abs. at 230 nm:0.20AU max
- UV Abs. at 250 nm:0.04AU max
- UV Abs. at 270 nm:0.01AU max
- UV Abs. at 300 nm:0.005AU max
- UV Cut-off:205nm max
- Water (H2O):0.04% max
Frequently Bought Together