ContactUsLinkComponent
AMSINO® Pre-Filled USP Sodium Chloride Injection Filled Syringe, 10 ml
AMSINO® Pre-Filled USP Sodium Chloride Injection Filled Syringe, 10 ml
Catalog # 77441-308
AMSINO® Pre-Filled USP Sodium Chloride Injection Filled Syringe, 10 ml
Catalog # 77441-308
Supplier Number:  IVF1010TM
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Sterility:
    Sterile
  • Volume:
    10 ml
  • Description:
    Pre-filled USP sodium chloride injection filled syringe
  • Cat. No.:
    77441-308
Specifications
About this item
Pre-filled syringe provides ready-to-use saline flush solution to help minimize the risk of catheter occlusion, improving patient safety.
  • Safety cap to avoid needlestick injuries
  • Compatible with needleless IV access system
  • Secure internal fluid sterilization
  • Large and suitable contact area for easy handling
Pre-filled syringes feature a distinct label design, with bold print and color-coded tip cap and label to allow easy indentification and decrease the risk of medication errors.
Packaging: Individually wrapped in sterile packaging.