Order Entry
United States
ContactUsLinkComponent
 
QuantiChrom™ α-Amylase Assay Kit
  77441-648
 :  
undefined
QuantiChrom™ α-Amylase Assay Kit
  77441-648
 :  DAMY2-100
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Description:
    QuantiChrom™ α-Amylase assay kit
  • Size:
    100 tests
  • Cat. No.:
    77441-648

 

 

For quantitative determination of α-amylase activity in biological samples.

  • Safe
  • Non-radioactive assay
  • Sensitive and accurate
  • Linear detection range of 0.2 to 100 U/L α-amylase in a 96-well plate assay
  • Convenient and high-throughput
  • Homogeneous 'mix-incubate-measure' type assay
  • Can be readily automated to assay thousands of samples per day

α-AMYLASE (EC 3.2.1.1) belongs to the family of glycoside hydrolase enzymes that break down starch into glucose molecules by acting on α-1,4-glycosidic bonds. The α-amylases cleave at random locations on the starch chain, ultimately yielding maltotriose and maltose, as well as glucose and 'limit dextrin' from amylose and amylopectin. In mammals, α-amylase is a major digestive enzyme. Increased α-amylase levels in humans are associated with salivary trauma, mumps due to inflammation of the salivary glands, pancreatitis and renal failure.

Simple, direct and automation-ready procedures for measuring amylase activity are very desirable. BioAssay Systems’ QuantiChrom™ α-Amylase Assay Kit provides a convenient colorimetric method to measure α-amylase activity in biological samples. In this assay, α-amylase hydrolyzes a synthetic substrate to release 2-chloro-4-nitrophenol. The rate of formation of the 2-chloro-4-nitrophenol, measured at 405 nm, is directly proportional to the enzyme activity.