Order Entry
United States
ContactUsLinkComponent
EnzyChrom™ Phosphoglucomutase Assay Kit
EnzyChrom™ Phosphoglucomutase Assay Kit
  77441-690
 :  
undefined
EnzyChrom™ Phosphoglucomutase Assay Kit
  77441-690
 :  EPGM-100
 :  

 

  • Description:
    EnzyChrom™ phosphoglucomutase assay kit
  • Size:
    100 tests
  • Cat. No.:
    77441-690

 

 

For quantitative determination of phosphoglucomutase enzyme activity in biological samples.

  • Safe and sensitive
  • Non-radioactive assay
  • Linear detection range in 96-well plate: 0.5 to 20 U/L activity
  • Fast and convenient
  • The procedure involves addition of a single working reagent and incubation for 20 min
  • Room temperature assay
  • No 37 °C incubator is needed
  • High-throughput
  • Homogeneous 'mix-incubate-measure' type assay
  • Can be readily automated to assay thousands of samples per day

Phosphoglucomutase (E.C. 5.4.2.2) is an enzyme which catalyzes the interconversion of glucose-1-phosphate (G1P) and glucose-6-phosate (G6P). Phosphoglucomutase (PGM) plays a crucial role in glycogen synthesis and degradation. PGM 1 deficiency can cause a rare congenital disorder of glycosylation. A decrease in PGM activity has also been linked with oxidative stress.

In BioAssay Systems EPGM-100 assay, PGM converts G1P to G6P, which is then oxidized by glucose-6-phosphate dehydrogenase. The formed NADPH is coupled to a formazan chromogen. The increase in absorbance at 460 nm is directly proportional to phosphoglucomutase activity.