ContactUsLinkComponent
DPD solution indicator, for residual chlorine analysis
DPD solution indicator, for residual chlorine analysis
Supplier:  Ricca Chemical
Certificates
undefined
DPD solution indicator, for residual chlorine analysis
Supplier:  Ricca Chemical
Certificates
Specification Test Results
  • Appearance:
    Clear, tan (preferred) to very light pink
  • Suitability for Use:
    Pink to red color in the presence of Chlorine