
Dithiothreitol (DTT, Cleland's reagent) ≥99.5%, white powder for electrophoresis
Catalog # IC856126
Supplier: MP Biomedicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle, amber
- Size:5 g
- Cat. No.:IC856126
- Supplier no.:856126
Specifications
Specification Test Results
- Identity Test:Passes
- Purity:≥99.5%
- FTIR:Conforms to Standard
Specification Test Results
About this item
:
:154.25 g/mol
Boiling Pt:? 125 °C (1013 hPa)
Melting Pt:40…50 °C
Flash Pt:≥ 110 °C ()
Storage Temperature:Refrigerator
EINECS:222-468-7
UN:3335
ADR:9,III
:GHS07
CAS Number:3483-12-3
:27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877