


N,N-Dimethylformamide ≥99.9%, OmniSolv® for biosynthesis, Supelco®
Catalog # EM-DX1732-1
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. No.:EM-DX1732-1
- Supplier no.:DX1732-1
Specifications
Specification Test Results
- Expiration date:Exp 24 months from mfg date
- Assay (GC):99.9% min
- Amines (as dimethylamine):5ppm max
- Color (APHA):10 max
- Filtered through 0.2 µm filter:To pass test
- Form:Clear liquid
- Infrared spectrum:Conforms to standard
- Residue after evaporation:5ppm max
- Titrable acid:0.4µeq/g max
- Titrable base:0.4µeq/g max
- UV Abs. at 270 nm:1.00AU max
- UV Abs. at 275 nm:0.30AU max
- UV Abs. at 280 nm:0.20AU max
- UV Abs. at 300 nm:0.05AU max
- UV Abs. at 350 nm:0.01AU max
- UV Abs. at 400 nm:0.005AU max
- UV Cut-off:270nm max
- Water (H2O):200ppm max
Specification Test Results
About this item
:
:73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–61 °C
Density:0.949 g/cm³ (20 °C)
Flash Pt:57.5 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
REACH:01-2119475605-32
:GHS02
:GHS08
:GHS07
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284