

N,N-Dimethylacetamide ≥99.9%, OmniSolv®, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:127-19-5
:
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686
:87.12 g/mol
Merck Index:13,03254
Boiling Pt:166.1 °C (1013 hPa)
Melting Pt:–20 °C
Density:0.937 g/cm³ (20 °C)
Flash Pt:70 °C
Storage Temperature:Ambient
EINECS:204-826-4
UN:2810
ADR:6.1,III
:GHS08
:GHS07
Specification Test Results
- Assay (GC):99.9% min
- Color (APHA):10 max
- Density at 25°C:0.931-0.941g/mL
- Filtered through 0.2 µm filter:To pass test
- Form:Clear liquid
- Infrared spectrum:Conforms to standard
- Residue after evaporation:5ppm max
- UV Abs. at 270 nm:1.00AU max
- UV Abs. at 280 nm:0.30AU max
- UV Abs. at 290 nm:0.15AU max
- UV Abs. at 310 nm:0.05AU max
- UV Abs. at 360 nm:0.01AU max
- UV Cut-off:270nm max
- Water (H2O):0.05% max
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_DX1543
SDS
MilliporeSigma The New M-Bottle for Solvents.pdf
Product Literature
Frequently Bought Together