Esteem® Stretchy Nitrile Gloves with Neu-Thera® Formulation, Cardinal
Supplier: Cardinal Health
Certificates
About this item
Special stretchy nitrile formulation provides the protection of nitrile while also offering comfort and flexibility.
- Thickness: 4.9 mil (finger)
Gloves are lined with the same ingredients found in physician-recommended hand lotions. Clinical tests have shown the Neu-Thera® formulation to moisturize and soothe skin through dermal conditioning during wear. Ambidextrous. Nonsterile. Powder- and latex-free. Fully textured.
Certifications: Gloves meet or exceed ASTM D3578 and pass the ASTM F1671 bacteriophage penetration test.
Specifications
- Catalog No:
- 89021-880
- Ambidextrous/Hand-specific:
- Ambidextrous
- Disposable/Reusable:
- Disposable
- Powdered/Powder-free:
- Powder-Free