


Copper(II) sulfate pentahydrate 98.5-100.5%, GenAR® USP for biotechnology, Macron Fine Chemicals™
Supplier: Avantor
MOC: Regulated Limited
About this item
CAS Number:7758-99-8
:
Formula:CuSO₄·5H₂O
MDL Number:MFCD00149681
:249.69 g/mol
Merck Index:13,02682
Density:2.286 g/cm³ (15.6 °C)
Storage Temperature:Ambient
EINECS:231-847-6
UN:3288
ADR:6.1,III
:Management of Change (MOC) category = RL
REACH:01-2119520566-40
:GHS05
:GHS09
:GHS08
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- USP - Assay (as CuSO₄):98.5 - 100.5 %
- USP - Calcium (Ca):≤0.005 %
- Solubility (5 in 50):Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Iron (Fe):≤0.003 %
- USP - Loss on Drying:33.0 - 36.5 %
- USP - Nickel (Ni):≤0.005 %
- USP - Potassium (K):≤0.01 %
- USP - Sodium (Na):≤0.02 %
- Endotoxin Concentration (EU/g):≤10
- EP/BP - Assay:99.0 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Chloride (Cl):≤100 ppm
- EP/BP - Iron (Fe):≤100 ppm
- EP/BP - Lead (Pb):≤50 ppm
- EP/BP - Loss on Drying:35.0 - 36.5 %
Specification Test Results
Related Information
Certificates
Search by Lot Number7790 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_MK779004 spec_MK779088
Product specification
Frequently Bought Together