

Copper(II) sulfate, anhydrous ≥98.5% (by iodometry), powder, BAKER ANALYZED®, J.T.Baker®
Catalog # JT1850-1
Supplier: Avantor
CAS Number:
MOC: Non-RegulatedCertificates
Specifications
- Pack type:Poly bottle
- Size:500 g
- Cat. No.:JT1850-1
- Supplier no.:1850-01
Specifications
Specification Test Results
- Assay (CuSO₄) (by iodometry):≥98.5 %
- Loss on Drying:≤1.0 %
- Calcium (Ca):≤0.005 %
- Iron (Fe):≤0.005 %
- Nickel (Ni):≤0.005 %
- Potassium (K):≤0.01 %
- Sodium (Na):≤0.02 %
- Lead (Pb):≤2.0 ppm
Specification Test Results
About this item
:
:159.61 g/mol
Merck Index:12,02722
Melting Pt:200 °C
Density:3.603 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-847-6
UN:3288
ADR:6.1,III
:Management of Change (MOC) category = N
:GHS07
:GHS09
CAS Number:7758-98-7
Formula:CuSO₄
MDL Number:MFCD00010981