Order Entry
United States
ContactUsLinkComponent
PYREX® Fernbach Culture Flask, Delong Neck, Corning
PYREX® Fernbach Culture Flask, Delong Neck, Corning
Catalog # 22877-172
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
PYREX® Fernbach Culture Flask, Delong Neck, Corning
Catalog # 22877-172
Supplier:  Corning
Supplier Number:  4424-2XL
Product available on GSA Advantage®
   Low Manufacturing Impact

Specifications

  • Capacity:
    2.8 L
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental Certification:
    I-RECs, ISO 14001 - Monterrey
  • Cat. No.:
    22877-172
  • Neck O.D.:
    38 mm
  • Supplier no.:
    4424-2XL

Specifications

About this item

Fernbach-style culture flask is ideal for culturing organisms that require a large surface area-to-volume ratio, and in serum production

  • Very good chemical resistance
  • Very good temperature resistance

Large marking spot allows for accurate labeling and easy identification. Flask bottom has three baffles at center on outside for maximal oxygen transfer to culture medium. Rimless, wide mouth, Delong-style neck is for use with plastic or stainless steel closures.

Caution: Do not use this flask on a heat source smaller than the base of the flask. Uneven heating of the flask could cause breakage.