Multi-Flex® Clean Class 100™ Latex Gloves, Cardinal
Supplier: Cardinal Health
Certificates
About this item
Powder-free latex gloves are clean-processed.
- Powder free, clean processed to protect sensitive environments
- Hand specific, tapered cuffs
- Anti-slip finish provides a sure grip
- Tapered cuffs
- Color: natural
Hand-specific gloves are tough, flexible, dependable, and strong. Thin fingertips ensure optimum fit, sensitivity, and dexterity required for small parts manufacturing procedures.
Ordering information: Multilevel bag-in-a-bag packaging, 50 pair per bag. Case totals reflect glove pairs (case of 400 includes 200 glove pairs).
Specifications
- Catalog No:
- PH2Y1605
- PH2Y1604
- PH2Y1601
- PH2Y1600
- PH2Y1603
- PH2Y1602
- PH2Y1607
- PH2Y1606
- Ambidextrous/Hand-specific:
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
Specifications
Related Information
Frequently Bought Together