Anti-H3K4un Rabbit Polyclonal Antibody
Catalog # 77437-824
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:H3K4un
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG (H, L)
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Gene ID:8350
- Storage Buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Storage Temperature:Store at –20 °C for 12 months.
- Concentration:1 µg/µl
- Purification:Purified by protein G
- Cat. No.:77437-824
- Packaging:Vial
Specifications
About this item
Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
Type: Primary
Antigen: H3K4un
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human