Anti-Wnt1 Rabbit Polyclonal Antibody
Catalog # 77440-014
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Wnt1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG (H, L)
- Reactivity:Human,Mouse
- Size:100 µl
- Gene ID:7471
- Storage Buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular Weight:39
- Storage Temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Cat. No.:77440-014
- Packaging:Vial
Specifications
About this item
Ligand for members of the frizzled family of seven transmembrane receptors. In some developmental processes, is also a ligand for the coreceptor RYK, thus triggering Wnt signaling. Probable developmental protein. May be a signaling molecule important in CNS development. Is likely to signal over only few cell diameters.
Type: Primary
Antigen: Wnt1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse