Anti-KCTD19 Rabbit Polyclonal Antibody
77438-096
: Bioss
- Antibody Type:Primary
- Antigen Name:KCTD19
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG (H, L)
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Gene ID:146212
- Storage Buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular Weight:105
- Storage Temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by Protein A
- Cat. No.:77438-096
- Packaging:Vial
KCTD19 potassium channel tetramerization domain containing 19.
Type: Primary
Antigen: KCTD19
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Mouse, Rat