Anti-CPNE2 Rabbit Polyclonal Antibody
Catalog # 77440-534
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:CPNE2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG (H, L)
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µl
- Gene ID:221184
- Storage Buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular Weight:61
- Storage Temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by Protein A
- Cat. No.:77440-534
- Packaging:Vial
Specifications
About this item
Calcium-dependent phospholipid-binding protein that plays a role in calcium-mediated intracellular processes. Exhibits calcium-dependent cell membrane binding properties.
Type: Primary
Antigen: CPNE2
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Mouse