


Acetic anhydride ≥97.0%, AR® ACS, Macron Fine Chemicals™
:108-24-7
:
:(CH₃CO)₂O
:MFCD00008705
:13,00057
:139.9 °C (1013 hPa)
:–73 °C
:1.08 g/cm³ (20 °C)
:49 °C
:Ambient
:203-564-8
:1715
:8,II
:Management of Change (MOC) category = N
:102.09 g/mol
:01-2119486470-36
:GHS02
:GHS05
:GHS07
- ACS - Assay ((CH₃CO)₂O):≥97.0 %
- Appearance (clear, colorless liquid):Passes Test
- ACS - Chloride (Cl):≤5 ppm
- ACS - Heavy Metals (as Pb):≤2 ppm
- ACS - Iron (Fe):≤5 ppm
- ACS - Phosphate (PO₄):≤0.001 %
- ACS - Residue after Evaporation:≤0.003 %
- ACS - Substances Reducing Permanganate:Passes Test
- ACS - Sulfate (SO₄):≤5 ppm
Frequently Bought Together