L(+)-Ascorbic acid ≥99.5%, powder, BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
CAS Number:50-81-7
:
Formula:C₆H₈O₆
MDL Number:MFCD00064328
:176.13 g/mol
Merck Index:13,00837
Boiling Pt:553 °C (1013 hPa)
Melting Pt:190…192 °C
Density:1.694 g/cm³ (20 °C)
:Management of Change (MOC) category = N
Storage Temperature:Ambient
EINECS:200-066-2
REACH:exempt
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Assay (C₆H₈O₆):≥99.5 %
- Specific Rotation, [Å]²⁵^D(c=10 in water)(+):20.5 - 21.5 Degree
- Residue after Ignition:≤0.10 %
- Loss on Drying at 105°C:≤0.1 %
- Heavy Metals (as Pb):≤0.001 %
- Trace Impurities - Iron (Fe):≤0.001 %
Specification Test Results
Related Information
Certificates
Search by Lot NumberB581 Product Specifications
Additional Product Details
sds_BAKRB581
SDS
Frequently Bought Together