Ammonia solution 30%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated, With Information
About this item
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Boiling Pt:32…37.7 °C (1013 hPa)
Melting Pt:–60…–57.5 °C
Density:0.88…0.91 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-647-6
ADR:8,III
:Management of Change (MOC) category = NI
UN:2672
:GHS05
:GHS07
:GHS09
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (as NH₃):28.0 - 30.0 %
- Specific Gravity at 60°/60°F:0.896 - 0.902
- Appearance:Passes Test
- Color (APHA):≤10
- Residue after Ignition:≤3 ppm
- Substances Reducing Permanganate:Passes Test
- Chloride (Cl):≤0.5 ppm
- Phosphate (PO₄):≤0.4 ppm
- Total Sulfur (as SO₄):≤1 ppm
- Trace Impurities - Aluminum (Al):≤20 ppb
- Trace Impurities - Arsenic and Antimony (as As):≤50.0 ppb
- Trace Impurities - Barium (Ba):≤10.0 ppb
- Trace Impurities - Boron (B):≤20.0 ppb
- Trace Impurities - Cadmium (Cd):≤5.0 ppb
- Trace Impurities - Calcium (Ca):≤200.0 ppb
- Trace Impurities - Chromium (Cr):≤5.0 ppb
- Trace Impurities - Cobalt (Co):≤1.0 ppb
- Trace Impurities - Copper (Cu):≤20.0 ppb
- Heavy Metals (as Pb):≤200.0 ppb
- Trace Impurities - Iron (Fe):≤10.0 ppb
- Trace Impurities - Lead (Pb):≤5.0 ppb
- Trace Impurities - Lithium (Li):≤20.0 ppb
- Trace Impurities - Magnesium (Mg):≤20 ppb
- Trace Impurities - Manganese (Mn):≤5.0 ppb
- Trace Impurities - Mercury (Hg):≤5.0 ppb
- Trace Impurities - Nickel (Ni):≤5.0 ppb
- Trace Impurities - Potassium (K):≤500.0 ppb
- Trace Impurities - Silicon (Si):≤1000.0 ppb
- Trace Impurities - Silver (Ag):≤5.0 ppb
- Trace Impurities - Sodium (Na):≤300 ppb
- Trace Impurities - Strontium (Sr):≤10.0 ppb
- Trace Impurities - Tin (Sn):≤10.0 ppb
- Trace Impurities - Zinc (Zn):≤5.0 ppb
Specification Test Results
Related Information
Certificates
Search by Lot Number28649125.pdf
SDS
9733 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together