Order Entry
United States
ContactUsLinkComponent
Ammonia solution 28.0 - 30.0 % (NH₃ basis), CMOS for microelectronic, J.T.Baker®
Corrosive
Irritant
Environment
Ammonia solution 28.0 - 30.0 % (NH₃ basis), CMOS for microelectronic, J.T.Baker®
MOC:  Regulated
Certificates
undefined
Corrosive
Irritant
Environment
Ammonia solution 28.0 - 30.0 % (NH₃ basis), CMOS for microelectronic, J.T.Baker®
MOC:  Regulated
Certificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Appearance:
    Passes Test
  • Assay (as NH₃):
    28.0 - 30.0 %
  • Color (APHA):
    ≤7
  • Insoluble Matter:
    ≤1 ppm
  • Residue after Ignition:
    ≤3 ppm
  • Carbon Dioxide (CO₂):
    ≤10 ppm
  • Pyridine:
    Passes Test
  • Substances Reducing Permanganate:
    Passes Test
  • Chloride (Cl):
    ≤0.3 ppm
  • Phosphate (PO₄):
    ≤0.20 ppm
  • Total Sulfur (as SO₄):
    ≤0.8 ppm
  • Trace Impurities - Aluminum (Al):
    ≤10.0 ppb
  • Trace Impurities - Arsenic (As):
    ≤50.0 ppb
  • Arsenic and Antimony (as As):
    ≤30.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤10.0 ppb
  • Trace Impurities - Beryllium (Be):
    ≤10.0 ppb
  • Trace Impurities - Bismuth (Bi):
    ≤20.0 ppb
  • Trace Impurities - Boron (B):
    ≤10.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤5.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤100.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤5.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤1.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤10.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤10.0 ppb
  • Trace Impurities - Germanium (Ge):
    ≤50.0 ppb
  • Trace Impurities - Gold (Au):
    ≤10.0 ppb
  • Heavy Metals (as Pb):
    ≤200.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤10.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤10.0 ppb
  • Trace Impurities - Lithium (Li):
    ≤10.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤50.0 ppb
  • Trace Impurities - Manganese (Mn):
    ≤5.0 ppb
  • Trace Impurities - Molybdenum (Mo):
    ≤10.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤5.0 ppb
  • Trace Impurities - Niobium (Nb):
    ≤10.0 ppb
  • Trace Impurities - Potassium (K):
    ≤100 ppb
  • Trace Impurities - Silicon (Si):
    ≤150.0 ppb
  • Trace Impurities - Silver (Ag):
    ≤5.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤100.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤10.0 ppb
  • Trace Impurities - Tantalum (Ta):
    ≤10.0 ppb
  • Trace Impurities - Thallium (Tl):
    ≤20.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤20.0 ppb
  • Trace Impurities - Titanium (Ti):
    ≤10.0 ppb
  • Trace Impurities - Vanadium (V):
    ≤10.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤5.0 ppb
  • Trace Impurities - Zirconium (Zr):
    ≤10.0 ppb
  • Particle Count - 0.5 µm and greater:
    ≤80 par/ml
  • Particle Count - 1.0 µm and greater:
    ≤10 par/ml

Specification Test Results

Related Information

Frequently Bought Together