Order Entry
United States
ContactUsLinkComponent
2-Nitro-m-xylene 99%
Irritant
2-Nitro-m-xylene 99%
Catalog # 77406-048
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
Irritant
2-Nitro-m-xylene 99%
Catalog # 77406-048
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  OR17602-250G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    250 g
  • Cat. No.:
    77406-048

Specifications

About this item

 :
 :151.16 g/mol
Boiling Pt:225…227 °C
Melting Pt:14…16 °C
Density:1.112
Flash Pt:88 °C
UN:1665
ADR:6.1,II
 :GHS07
CAS Number:81-20-9
Formula:C₈H₉NO₂
MDL Number:MFCD00007181