

Ammonium heptamolybdate tetrahydrate (81.0 - 83.0% MoO₃), crystals, BAKER ANANYZED™ ACS
Supplier: Avantor
MOC: Non-Regulated
About this item
CAS Number:12054-85-2
:12027-67-7
Formula:(NH₄)₆Mo₇O₂₄·4H₂O
MDL Number:MFCD00167059
:1235.86 g/mol
Merck Index:13,00536
Density:2.50 g/cm³ (20 °C)
:Management of Change (MOC) category = N
EINECS:234-722-4
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Assay (as MoO₃):81.0 - 83.0 %
- Insoluble Matter:≤0.005 %
- Chloride (Cl):≤0.002 %
- Nitrate (NO₃):Passes Test
- Arsenate, Phosphate and Silicate (as SiO₂):≤0.001 %
- ACS - Phosphate (PO₄):≤5 ppm
- Sulfate (SO₄):≤0.02 %
- Heavy Metals (as Pb):≤0.001 %
- Magnesium (Mg):≤0.005 %
- Potassium (K):≤0.01 %
- Sodium (Na):≤0.010 %
Specification Test Results
Related Information
Certificates
Search by Lot Number0716 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Brand Definition Baker Analyzed
Datasheet
Frequently Bought Together