Standard Exposure Xylene and Chemical-Resistant Labels
Supplier: GA INTERNATIONAL INC
Certificates
About this item
Permanent chemical-resistant labels are intended for identifying glass microscope slides used in histology applications. They withstand exposure and immersion in harsh solvents, such as xylene and newer xylene alternatives/substitutes. Perfect for use in various staining protocols (routine and special).
- Temperature range: –80 to 120 °C (–112 to 248 °F)
- Superior resistance against harsh chemicals and solvents
- Chemical-resistant printout will not smear or smudge
- Adhesive ensures permanent secure adhesion to container surface
- Stain-resistant to Hematoxylin and Eosin, as well as other typical histology dyes
- Printable with all major thermal transfer printer brands
- Recommended ink ribbon: XAR-Class
Chemical-resistant labels designed to fit microscope slides, tissue cassettes, bottles, or containers exposed or immersed in xylene and other harsh solvents, alcohols, and stains. Perfect for use in histology and chemistry labs. Labels will adhere permanently from –80 ˚C (–112 ˚F) to 120 ˚C (248 ˚F), and can be safely used in ovens, boiling water, and microwaves.
Labels can be printed with all major brands of thermal transfer printers.
Frequently Bought Together