


Acetic anhydride ≥98%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
CAS Number:108-24-7
:
Formula:(CH₃CO)₂O
MDL Number:MFCD00008705
Merck Index:13,00057
Boiling Pt:139.9 °C (1013 hPa)
Melting Pt:–73 °C
Density:1.08 g/cm³ (20 °C)
Flash Pt:49 °C
Storage Temperature:Ambient
EINECS:203-564-8
UN:1715
ADR:8, 3,II
:Management of Change (MOC) category = N
:102.09 g/mol
REACH:01-2119486470-36
:GHS02
:GHS05
:GHS07
Specification Test Results
- Assay ((CH₃CO)₂O):≥98 %
- Color (APHA):≤10
- Residue after Evaporation:≤0.0020 %
- Substances Reducing Permanganate:Passes Test
- Chloride (Cl):≤3 ppm
- Phosphate (PO₄):≤5 ppm
- ACS - Sulfate (SO₄):≤5 ppm
- Heavy Metals (as Pb):≤2 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number0018 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Brand Definition Baker Analyzed
Datasheet
spec_JT0018-3
Product specification
Frequently Bought Together