ContactUsLinkComponent
PYREX® Graduates, Beakers Form, Double Spout, Double Scale
PYREX® Graduates, Beakers Form, Double Spout, Double Scale
Catalog # 89090-716
Supplier:  Corning
   Sustainable Options Available
Supplier #: 6480-1L
Product available on GSA Advantage®
Low Manufacturing Impact
undefined
PYREX® Graduates, Beakers Form, Double Spout, Double Scale
Catalog # 89090-716
Supplier:  Corning
Supplier Number:  6480-125
   Sustainable Options Available
Supplier #: 6480-1L
Product available on GSA Advantage®
  Low Manufacturing Impact
Log in for availability
Select an Option
50 - 1000 ml
Select an Option
1 L (34 oz.)
Select an Option
89090-716
Select an Option
6480-1L
Specifications
  • Catalog No:
  • 89090-716
  • 89091-246
  • 89091-130
  • 89090-850
  • 89091-406
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental Certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Specifications
About this item
PYREX®, borosilicate glass, double spout, double scale beakers.
  • Increased wall thickness for better mechanical properties
  • Ideal for measuring hot solutions
  • Should not have direct contact with heat source
Thick walled, with spout, approximate graduations for both fluid ounces and milliliters.
Frequently Bought Together