Protektive Pak® ESD Stacking Board Handling Trays, Desco Industries
Supplier: Protektive Pak
Certificates
About this item
Provide complete ESD and physical protection for static-sensitive circuit boards and eliminate the need for non-recyclable static shielding bags
Constructed with double sides, triple ends, and a double thickness bottom, these durable handlers minimize rapid discharge and sparking. Feature a static-dissipative surface of 107–109ohms/sq.
Frequently Bought Together