PCR Rack, 32-Place
Blue, 32, 0.2 ml tubes, PP
Catalog # 77395-604
Supplier: Heathrow Scientific
Supplier #: 120889
Jump to:
Product Details & Documents
Efficient PCR sampling and storage.
- Small and compact design is stackable, saving valuable space in the freezer
- Right sized when processing smaller batches
- Hinged lid allows lid to to sit at 180 degrees, clear of sample processing, or lid can be removed entirely
- Lightweight and durable construction for reusability
Designed for pre and post-PCR sampling. Streamline PCR workflows with this 32-place PCR rack. This versatile rack accommodates 0.2 ml tubes both individually and in strips of 8, as well as 32-place assay plates (8×4).
The transparent lid protects samples and allows for quick assessment of contents. Embossed numbers and letters on the wells provide easy identification and tracking of samples.
Specifications
- No. of Holes:32
- Color:Blue
- Material:PP
- For use with:0.2 ml tubes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



