Nalgene® Lowboy™ Carboys with Spigot, High-Density Polyethylene, Thermo Scientific
Catalog # 16117-969
Supplier: Thermo Fisher Scientific
Specifications
- Capacity:8 L (2.1 gal.)
- Dimensions:16.5 x 36.8 x 30.5 cm (61/2 x 141/2 x 12")
- Environmentally Preferable:Low Manufacturing Impact
- Environmental Certification:ISO14001 (Certificate Registration No. EMS 752603)
- Cat. No.:16117-969
- Supplier no.:2323-0008
Specifications
About this item
Ideal for storing and dispensing reagents, titrants, and distilled water.
- Leakproof
For replacement caps, see cap 16219-104 in 16219-030 series.
Tolerates strong acids and bases. Not recommended for use with aggressive organic solvents. Low-profile design fits easily in tight spaces, on shelves, or in refrigerators. Can be stacked three high. Convenient integral handgrip for safe, easy transport. Cap Size: 63-415.
Ordering information: Supplied with carboy, polypropylene spigot, and screw cap.