Order Entry
United States
ContactUsLinkComponent
Halobacterium salinarum S9 Native Bacteriorhodopsin
Halobacterium salinarum S9 Native Bacteriorhodopsin
Catalog # 77392-866
Supplier:  CUBE BIOTECH
CAS Number:  
undefined
Halobacterium salinarum S9 Native Bacteriorhodopsin
Catalog # 77392-866
Supplier:  CUBE BIOTECH
Supplier Number:  28903
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Native
  • Species:
    Halobacterium salinarum S9
  • Size:
    500 µg
  • Amino Acid Number:
    262
  • Protein Synonyms:
    BR, HsBR, Bacteriopsin
  • Protein/Peptide Name:
    Bacteriorhodopsin
  • Purity:
    >95%, determined via SDS-PAGE
  • Molecular Weight:
    28256 Da
  • Sequence:
    MLELLPTAVE GVSQAQITGR PEWIWLALGT ALMGLGTLYF LVKGMGVSDP DAKKFYAITT LVPAIAFTMY LSMLLGYGLT MVPFGGEQNP IYWARYADWL FTTPLLLLDL ALLVDADQGT ILALVGADGI MIGTGLVGAL TKVYSYRFVW WAISTAAMLY ILYVLFFGFT SKAESMRPEV ASTFKVLRNV TVVLWSAYPV VWLIGSEGAG IVPLNIETLL FMVLDVSAKV GFGLILLRSR AIFGEAEAPE PSAGDGAAAT SD
  • Cat. No.:
    77392-866

Specifications

About this item

Bacteriorhodopsin (BR) is a 7-transmembrane protein that acts as a light-driven proton pump in archaebacteria.

  • Alternative names: Bacteriorhodopsin, BR, HsBR, Bacteriopsin
  • UniProt Number: P02945
  • Protein class: 7-transmembrane protein
  • Organism: Halobacterium salinarum S9
  • Sequence, explained: Full-length, wildtype sequence with N-terminal signal sequence which is mostly absent in the preparation
  • Affinity tags: None
  • Expression host: Halobacterium salinarum S9
  • Buffer composition: Purple membrane (PM) patches in H₂O, 0.02% NaN₃
  • Purified via: Sequential centrifugation
  • Absorbance: Extinction coefficient at 560 nm: 63000 M⁻¹cm⁻¹
  • Activity: Binding of ligand all-trans retinal, covalently bound to a lysine residue
  • Evaluation of UV-VIS spectrum: absorbance ratio 280/560 nm = 2.3
  • Function: Photoreceptor protein undergoing a photocycle, Retinal protein, light-driven proton pump

Due to BR's covalently bound colored ligand, BR is widely used as a control membrane protein, for example for crystallization assays and biochemical/biophysical experiments. We provide BR of high purity, prepared as purple membrane patches from Halobacterium salinarum.