SARS-CoV-2 Recombinant SARS CoV-2 Spike Protein Set in LMNG Detergent, DIBMA Glycerol, Nanodisc Complex (from HEK293 cells)
Catalog # 77392-856
Supplier: Cube Biotech
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:SARS-CoV-2
- Size:100 µg
- Storage Conditions:−80 °C, avoid freeze-thaw cycles
- Amino Acid Number:1286 (Including Rho1D4-tag and linker)
- Protein Synonyms:Spike glycoprotein
- Protein/Peptide Name:SARS CoV-2 Spike Protein Set in LMNG Detergent, DIBMA Glycerol, Nanodisc Complex
- Purity:>98%, determined via SDS-PAGE
- Molecular Weight:142114 Da
- Shipping Temperature:Dry Ice
- Cat. No.:77392-856
Specifications
About this item
The coronavirus, also known as SARS-CoV-2, enters the cell by using its surface SPIKE. SPIKE is processed on the cell's surface by TMPRSS2, a serine protease. It then subsequently binds to ACE2 a cell surface receptor.
- Alternative names: SPIKE_SARS2 Spike glycoprotein
- UniProt Number: P0DTC2
- Protein class: Single span transmembrane protein
- Organism: Severe acute respiratory syndrome coronavirus 2 (2019-nCoV) (SARS-CoV-2)
- Sequence, explained: Full-length sequence (aa 1 - 1273), furin cleavage site 'RRAR' mutated to 'GSAG'; C-terminal Rho1D4 tag fused with spacer 'GSSG' to protein sequence
- Affinity tags: C-terminal Rho1D4-tag
- Expression host: Hek293 Expi cells
- Buffer composition: 20 mM Hepes pH 7.5; 150 mM NaCl, 0.01 % LMNG
- Purified via: PureCube Rho1D4 Agarose
- Function: Host cell surface receptor binding; fusion of virus membrane with host endosome membrane
The Native SPIKE protein is a trimer that is located in the coronavirus membrane. Therefore to get pure and native SPIKE this trimer needs to be kept intact. And Cube Biotech offers stabilized intact form of SPIKE.