Order Entry
United States
ContactUsLinkComponent
Human Recombinant MSP2N2-D73C (from E. coli)
Human Recombinant MSP2N2-D73C (from E. coli)
Supplier:  CUBE BIOTECH
Certificates
undefined
Human Recombinant MSP2N2-D73C (from E. coli)
Supplier:  CUBE BIOTECH

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Amino Acid Number:
    392
  • Protein Synonyms:
    Membrane Scaffold Protein 1D1 D73C
  • Protein/Peptide Name:
    MSP2N2-D73C
  • Purity:
    >90% (Determined by SDS-PAGE)
  • Molecular Weight:
    44661 Da
  • Sequence:
    MGHHHHHHH DYDIPTT ENLYFQG STFSKLREQLG PVTQEFWCNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ GT PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ

Specifications

Related Information

Frequently Bought Together