Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant MSP2N2 (from E. coli)
Mouse Recombinant MSP2N2 (from E. coli)
Supplier:  Cube Biotech
Certificates
undefined
Mouse Recombinant MSP2N2 (from E. coli)
Supplier:  Cube Biotech

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Amino Acid Number:
    371 (Human origin)
  • Protein Synonyms:
    Membrane Scaffold Protein 2N2
  • Protein/Peptide Name:
    MSP2N2
  • Purity:
    >90% (Determined by SDS-PAGE)
  • Molecular Weight:
    4305 kDa (Human origin)
  • Sequence:
    G STFSKLREQLG PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ GT PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ

Specifications

Related Information

Frequently Bought Together