Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant MSP1E3D1 (from E. coli)
Mouse Recombinant MSP1E3D1 (from E. coli)
Supplier:  CUBE BIOTECH
Certificates
undefined
Mouse Recombinant MSP1E3D1 (from E. coli)
Supplier:  CUBE BIOTECH

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Amino Acid Number:
    256 (Human origin), 254 (Mouse origin)
  • Protein Synonyms:
    Membrane Scaffold Protein 1E3D1
  • Protein/Peptide Name:
    MSP1E3D1
  • Purity:
    >90% (Determined by SDS-PAGE)
  • Molecular Weight:
    29982 Da (Human origin), 29786 (Mouse origin)
  • Sequence:
    G STFSKLREQLG PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ

Specifications

Related Information

Frequently Bought Together