Human Recombinant MSP1E3D1 (from E. coli)
Supplier: Cube Biotech
Certificates
About this item
Membrane scaffold protein, human origin.
- Extinction coefficient (in water) ε280: 26930 M⁻¹cm⁻¹ (Human origin), 29450 M⁻¹cm⁻¹ (Mouse origin)
- Buffer: 20 mM TRIS pH 7.4; 100 mM NaCl; 0.5 mM EDTA
- Affinity tag: none
Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1E3D1 creates nanodiscs of 12 to 14 nm in diameter.
This product is available with different species as its background. This is important for e.g. for secondary antibodies of a western blot.
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:Storage of lyophilized copolymer: −20 °C for several years; Storage of reconstituted protein: 2...8 °C for several day
- Amino Acid Number:256 (Human origin), 254 (Mouse origin)
- Protein Synonyms:Membrane Scaffold Protein 1E3D1
- Protein/Peptide Name:MSP1E3D1
- Purity:>90% (Determined by SDS-PAGE)
- Molecular Weight:29982 Da (Human origin), 29786 (Mouse origin)
- Sequence:G STFSKLREQLG PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ
- Shipping Temperature:RT
Specifications
Frequently Bought Together