Cardboard Slide Tray, 20-Place
Supplier: Heathrow Scientific
Certificates
About this item
Cardboard slide trays hold standard slides (76×26 mm and 3×1").
- Organize and protect
- Index on the front cover corresponds with the number grid inside for identification
- Recessed slide compartments and inside covers for specimen protection
- Slides cannot be accessed when cover is closed reducing contamination
- Sturdy compartment holder is made from a double layer of densely pressed cardboard
- Cover is single layer cardboard
- Front protection flaps are attached with durable tape
The cardboard slide tray offers a convenient solution for working safely, transporting, and short-term storage of slides.
Frequently Bought Together