ContactUsLinkComponent
Vinyl-Coated Lead Rings 'C' Shape, Blue
Vinyl-Coated Lead Rings 'C' Shape, Blue
Catalog # 77395-448
undefined
Vinyl-Coated Lead Rings 'C' Shape, Blue
Catalog # 77395-448
Supplier Number:  HS8881D
Specifications
  • Description:
    Vinyl-coated lead rings ('C' shape), blue
  • Cat. No.:
    77395-448
Specifications
About this item
Stabilize vessels and labware on the bench or in water baths.
  • Vinyl-coated lead ring will not mark or scratch benchtops and labware
  • C-shape style allows some manual size variation for a customized fit
  • Fits 1000 to 4000 ml
The lead ring is perfect for stabilizing vessels and labware or for water bath immersions.