Order Entry
United States
ContactUsLinkComponent
Anti-HighSpec Rho1D4 antibody
Anti-HighSpec Rho1D4 antibody
Catalog # 77393-144
Supplier:  CUBE BIOTECH
undefined
Anti-HighSpec Rho1D4 antibody
Catalog # 77393-144
Supplier:  CUBE BIOTECH
Supplier Number:  40020

Specifications

  • Size:
    200 µg
  • Cat. No.:
    77393-144

Specifications

About this item

This is an antibody specifically for targeting a Rho1D4-(TETSQVAPA)-tag on a protein.

  • Antibody name: Rho-1d4-1
  • Cell line/origin: Mouse, Hybridoma
  • Antibody purification: rProtein A
  • Quantity: 200 µg
  • Concentration: 4.11 mg/ml (by UV absorbance at 280 nm)
  • Physical state: Liquid (sterile filtered)
  • Color: Clear
  • Buffer: 0.25 M dodium bicarbonate, 0.5 M sodium chloride, pH=8.3
  • Purity: >95% (determined by non-denaturing SDS-PAGE)
  • Shipment temperature: Shipped on ice
  • Short-term storage: Store vial at 4 °C. Centrifuge product if not completely clear after standing at room temperature and/or heating to 37 °C
  • Long-term storage: Alinquot contents and freeze at −20 °C or below. Avoid repeated freeze-thaw cycles

The Rho1D4-antibody can be used in Western Blots and ELISAs. Its origin is mouse and should be stored at 20 °C for the long-term.

Type: Recombinant
Antigen: HighSpec Rho1D4 antibody
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity