Order Entry
United States
ContactUsLinkComponent
 
HighSpec Rho Starter Set 3: HighSpec Compact Cartridge Rho1D4 + Rho1D4 Peptide
  77393-088
 :  
undefined
HighSpec Rho Starter Set 3: HighSpec Compact Cartridge Rho1D4 + Rho1D4 Peptide
  77393-088
 :  33398
 :  

 

  • Size:
    1 ml + 5 mg
  • Cat. No.:
    77393-088

 

 

Our Rho1D4 starter set 3 combines our HighSpec Rho1D4 compact cartridge and the Rho1D4 elution peptide.

  • Usage: As a matrix for Hydrophobic Interaction Chromatography (HIC)
  • Specificity: Hydrophobic biomolecules and proteins
  • Bead size: 40 μm
  • Bead ligand: Butyl groups
  • Butyl group density: 40 µmol/ml
  • BSA binding capacity: 25 mg/ml
  • Lysozyme binding capacity: 45 mg/ml
  • pH stability: 2 to 13
  • Filling quantity: Delivered as a 50% suspension
  • Matrix: 7.5% cross-linked agarose
  • Short term storage: In neutral buffer at 4 °C
  • Long term storage: 20 mM sodium acetate, 20% ethanol, pH 6.5 at 4 °C
  • Chemical structure of butyl ligand: Butyl group structure

The compact cartridge is an FPLC column pre-packed with our HighSpec Rho1D4 agarose resin. It is meant for protein purification assays of the highest possible purity, exploiting the specificity of an antibody to its antigen.