Acetic acid glacial 99.7-100.5%, ARISTAR® ACS, meets analytical specification of USP, FCC, VWR Chemicals BDH®
Supplier: BDH
Certificates
About this item
Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
Boiling Pt:118
Melting Pt:17
Density:1.05 g/cm³ (20 °C)
Molecular Weight:60.05 g/mol
Flash Pt:38.5 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8 II
REACH:01-2119475328-30
:GHS02
:GHS05
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (CH3COOH):99.7% min.(ACS)/99.5-100.5%(USP/FCC)
- Acetic Anhydride:0.01% max.(ACS)
- Chloride (as Cl):To Pass Test(USP)
- Chloride (Cl):1 ppm max.(ACS)
- Color (APHA):10 max.(ACS)
- Congealing Temperature:15.6 C min.(USP/FCC)
- Dilution Test:To pass test(ACS)
- Heavy Metals (as Pb):0.5 ppm max.(ACS)
- Identification::acetate To Pass Test(USP/FCC)
- Iron (Fe):0.2 ppm max.(ACS)
- Lead (Pb):0.5 mg/kg max.(FCC)
- Nonvolatile residue:1.0mg max.(USP)/0.005% max.(FCC)
- Oxidizable substances:To pass test(USP/FCC)
- Residue After Evaporation:0.001% max.(ACS)
- Substances Reducing Dichromate:To pass test (ACS)
- Substances Reducing Permanganate:To pass test (ACS)
- Sulfate:1 ppm max.(ACS)
- Sulfate (SO4):To Pass Test(USP)
- Titrable Base:0.0004 meq/g max.(ACS)
- Manufacture Date:Record Actual
- Expiration date:Record Actual
Specification Test Results
Related Information
Certificates
Search by Lot Number5 Gal (19 l) Poly Vented Flow Carboy Packaging
Packaging Information
Avantor Portfolio of Chemicals Avantor Top Rated Guide BDH ACS Acids in PPQ Bottles Flyer VWR® Chemicals in Poly-Coated Glass Bottles
Product Literature
BDH3092-500MLP_P BDH3094-2.5LG_P BDH3096-2.5LPC_P BDH3098-3.8LP_P BDH3100-19L_P
BSE/TSE Certificates
More
Freezing Acetic BDH VWR GHS Compliance Letter (US)
Additional Product Details
spec_BDHVBDH3092
Product specification
Frequently Bought Together