ContactUsLinkComponent
Mini Nutating Rocker, 230 V
  470107-128
 :  Ward's
Irregular Voltage
undefined
Mini Nutating Rocker, 230 V
  470107-128
 :  Ward's
 :  H3D-1020-E
Irregular Voltage
  • Cat. No.:
    470107-128
  • Electrical:
    230 V
  • Supplier no.:
    H3D-1020-E
This mini mixer takes up minimal bench space, while providing maximum mixing efficiency.
  • 230 V model
  • Fixed speed
  • Small platform
  • Flat cord stays out of your way
  • Two year warranty
Capable of supporting loads of up to four pounds, this fixed speed orbital mixer is small but powerful. With 3D nutating movement, it will agitate your samples gently, but thoroughly.
 : 230 V model. Corresponding model with voltage suitable for use in US (470107-056) is also available, please see separate product page.