Order Entry
United States
ContactUsLinkComponent
Thioanisole 99%
Flammable
Irritant
Thioanisole 99%
Catalog # 77407-556
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
Flammable
Irritant
Thioanisole 99%
Catalog # 77407-556
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  OR2490-5G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    5 g
  • Cat. No.:
    77407-556

Specifications

About this item

 :
 :124.21 g/mol
Boiling Pt:188 °C
Melting Pt:–15 °C
Density:1.057 g/cm³ (25 °C)
Flash Pt:57 °C
Storage Temperature:Ambient
EINECS:202-878-2
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:100-68-5
Formula:C₇H₈S
MDL Number:MFCD00008559