Order Entry
United States
ContactUsLinkComponent
Tetrafluoroboric acid min 48% in aqueous solution
Corrosive
Toxic
Tetrafluoroboric acid min 48% in aqueous solution
Catalog # 77414-850
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
Corrosive
Toxic
Tetrafluoroboric acid min 48% in aqueous solution
Catalog # 77414-850
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  PC3728-500G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    500 g
  • Cat. No.:
    77414-850

Specifications

About this item

 :
 :87.81
Boiling Pt:130 °C
Density:1.41 g/cm³ (25 °C)
Flash Pt:66 °C
UN:1775
ADR:8,II
 :GHS05
 :GHS06
Storage Temperature:Ambient
CAS Number:16872-11-0
Formula:HBF₄
MDL Number:MFCD00011345