Order Entry
United States
ContactUsLinkComponent
(6-Oxo-1,6-dihydropyridin-3-yl)boronic acid 97%
Toxic
(6-Oxo-1,6-dihydropyridin-3-yl)boronic acid 97%
Catalog # 77409-126
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
Toxic
(6-Oxo-1,6-dihydropyridin-3-yl)boronic acid 97%
Catalog # 77409-126
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  OR321272-250MG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    250 mg
  • Cat. No.:
    77409-126

Specifications

About this item

 :
 :138.917 g/mol
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:903899-13-8
Formula:C₅H₆BNO₃
MDL Number:MFCD22123505