Order Entry
United States
ContactUsLinkComponent
Methanol-[D₄] (>99,8% D) (max. 0.03% TMS (tetramethylsilan))
Flammable
Toxic
Health
Methanol-[D₄] (>99,8% D) (max. 0.03% TMS (tetramethylsilan))
Catalog # 77401-276
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
Flammable
Toxic
Health
Methanol-[D₄] (>99,8% D) (max. 0.03% TMS (tetramethylsilan))
Catalog # 77401-276
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  DE70FT-10ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Septum vials
  • Size:
    10 ml
  • Cat. No.:
    77401-276

Specifications

About this item

 :
 :36.01 g/mol
Boiling Pt:65.4 °C
Melting Pt:–99 °C
Density:0.888 g/cm³ (25 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:212-378-6
UN:1230
ADR:3+6.1,II
 :GHS02
 :GHS06
 :GHS08
CAS Number:811-98-3
Formula:D₃COD
MDL Number:MFCD00044637