Order Entry
United States
ContactUsLinkComponent
Aquatic Block™ Serum-Free PBS (with Surfactant)
Aquatic Block™ Serum-Free PBS (with Surfactant)
Catalog # 77395-190
CAS Number:  
undefined
Aquatic Block™ Serum-Free PBS (with Surfactant)
Catalog # 77395-190
Supplier Number:  PP83A-500
CAS Number:  

Specifications

  • Description:
    Aquatic Block™ Serum-free PBS (with surfactant), 500 ml
  • Cat. No.:
    77395-190

Specifications

About this item

A non-mammalian blocking reagent supplied in PBS for diagnostic systems involving membrane or other solid phase material.

  • Imparts superior stability and flow characteristics
  • Utilizes proteins that do not interact with mammalian antibodies
  • Supplied in HDPE plastic

Preservative - 0.09% sodium azide, with surfactant.

Applications: ELISA/EIA/Western blot.