About this item
Highest quality FBS, HyClone™ AU-origin defined FBS is proprietary serial 40 nm-filtered, virus-tested to 9 CFR standard, and supplied with an extensive biochemical profile.
- Well-defined: assay results for over 50 components, with additional testing available
- AU origin: sourced in Australia and processed in New Zealand with complete traceability to origin
- Ultra-low endotoxin: ≤5 EU/ml endotoxin for reduced research and production risk
- Sterile-filtered: proprietary serial 40 nm filter process
γ-irradiated and heat-inactivated FBS options also available.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77395-500
- 77395-496
- 77395-498
- 77395-492
- 77395-494
- 77395-488
- 77395-490
- 77395-502
- Origin:
- Animal
- Animal
- Animal
- Animal
- Animal
- Animal
- Animal
- Animal
- Endotoxin:
- ≤5 EU/ml
- ≤5 EU/ml
- ≤5 EU/ml
- ≤5 EU/ml
- ≤5 EU/ml
- ≤5 EU/ml
- ≤5 EU/ml
- ≤5 EU/ml
- Hemoglobin:
- ≤15 mg/dl
- ≤15 mg/dl
- ≤15 mg/dl
- ≤15 mg/dl
- ≤15 mg/dl
- ≤15 mg/dl
- ≤15 mg/dl
- ≤15 mg/dl
- Storage Temperature:
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.