ContactUsLinkComponent
Cap2™ PCR Tubes, 0.2 ml Dual-Cap Sample Collection
Cap2™ PCR Tubes, 0.2 ml Dual-Cap Sample Collection
Catalog # 77321-968
Supplier:  AZENTA US, INC
Cap2™ PCR Tubes, 0.2 ml Dual-Cap Sample Collection
Catalog # 77321-968
Supplier:  AZENTA US, INC
Supplier Number:  4TI-0791
Specifications
  • Description:
    Cap2™ 0.2 ml dual-cap sample collection PCR tubes
  • Cat. No.:
    77321-968
Specifications
About this item
A unique PCR tube featuring a patent pending dual-cap mechanism, 2D datamatrix code as well as a human readable identification, allowing for wide-ranging workflow versatility. Innovative features allow for utilization as a consumable suitable for sample collection/transport that can be directly processed for analysis, ideal for genetic testing workflows.
  • Patent pending dual-cap design with both hinged lid and screw cap
  • Sample identification from 2D code and human readable number
  • Tab on side of cap includes a 2D datamatrix code and human readable number
  • Tab on collar of hinged lid makes for quick and secure sample tracking
  • Capability to work with automation, tubes are compatible with 48-format SBS footprint rack
  • PCR tube with patent pending cap-in-cap design, sample identifiers and automation friendly capabilities offer a flexible, scalable solution
  • Free from DNase, RNase, human genomic DNA, and endotoxin/pyrogen
  • Leak tested
  • Polypropylene, ultra-thin walled, low DNA binding tubes make for optimum PCR results and mitigation against DNA loss from samples
  • 0.2 ml (200 µl) total well capacity
The novel cap-in-cap design features a hinged lid and automation friendly screw cap, offering flexibility for use in manual workflows, alongside mitigating against contamination, as well as workflow scale up capabilities and consumable consolidation. This unique solution to error-prone, slow and labor-intensive workflows offers a flexible, scalable solution for PCR and genetic testing applications.
SBS rack
Product Information