ContactUsLinkComponent
Glass Cutter, Natural Diamond, Kemtech America
Glass Cutter, Natural Diamond, Kemtech America
Catalog # 60004-970
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Glass Cutter, Natural Diamond, Kemtech America
Catalog # 60004-970
Supplier:  Kemtech America
Supplier Number:  DC18001
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Description:
    Glass Cutter
  • Cat. No.:
    60004-970
  • Supplier no.:
    DC18001
Specifications
About this item
This cutter is designed with rack on shank for breaking off single- or double-strength glass.
  • Extra-large natural diamond for durability
Handheld plate glass cutter has a durable diamond tip for extended life.